Learn More
Ubpbio PROTEIN A/G AGAROSE, 1ML
Catalog No. 501126960
Encompass_Preferred
Protein Purification Reagents – Protein A/G Resin (UBPBio catalog# P5030-1, P5030-5, P5030-25) – Protein A/G XPure Agarose Resin consists of recombinant protein A and protein G covalently bound to crosslinked agarose beads, which minimizes leakage of Protein A/G and allow reuse the resin. The protein A/G resin has a binding capacity of ~ 20 mg human IgG/ml resin and works well in batch or FPLC column purifications.
Catalog No. 50-112-6960 Supplier Ubpbio Supplier No. P50301
Protein Purification Reagents – Protein A/G Resin (UBPBio catalog# P5030-1, P5030-5, P5030-25) – Protein A/G XPure Agarose Resin consists of recombinant protein A and protein G covalently bound to crosslinked agarose beads, which minimizes leakage of Protein A/G and allow reuse the resin. The protein A/G resin has a binding capacity of ~ 20 mg human IgG/ml resin and works well in batch or FPLC column purifications.