Learn More
Genscript Corporation Protein A/G Magbeads MX
Catalog No. 5000058067
Encompass
The GenScript Protein A/G MagBeads MX are ideal for small scale purification of antibodies from plasma serum ascites and cell culture
Catalog No. 50-000-58067 Supplier Genscript Corporation Supplier No. L008942
The GenScript Protein A/G MagBeads MX are ideal for small scale purification of antibodies from plasma serum ascites and cell culture