Learn More
Genscript Corporation Protein A/G Magbeads MX
Supplier:  Genscript Corporation L0089450
Encompass
The GenScript Protein A/G MagBeads MX are ideal for small scale purification of antibodies from plasma serum ascites and cell culture
Catalog No. 50-000-58068