Learn More
Genscript Corporation Protein A/G MagBeads
Catalog No. 5000058363
Encompass
GenScript Protein A/G MagBeads are superparamagnetic beads covalently bound with recombinant proteins A and G for antibody purification The beads are supplied as 25% slurry in phosphate buffered saline (PBS) pH 7 4 containing 20% ethanol The Protein A/G MagBeads have a binding capacity of more than 10 mg Goat IgG per 1 ml settled beads
Catalog No. 50-000-58363 Supplier Genscript Corporation Supplier No. L00277
GenScript Protein A/G MagBeads are superparamagnetic beads covalently bound with recombinant proteins A and G for antibody purification The beads are supplied as 25% slurry in phosphate buffered saline (PBS) pH 7 4 containing 20% ethanol The Protein A/G MagBeads have a binding capacity of more than 10 mg Goat IgG per 1 ml settled beads