Learn More
Medchemexpress LLC HY-K0202 5 mL Medchemexpress, Protein A/G Magnetic Beads CAS: Purity:>98%
SDP
Catalog No. 501933300
Encompass_Preferred
Medchemexpress, HY-K0202 5 mL Protein A/G Magnetic Beads CAS: Protein A/G Magnetic Beads provide a fast and convenient method for Immunuoprecipitaion and Co-Immunoprecipitation and Chromatin Immunoprecipitation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3300 Supplier Medchemexpress LLC Supplier No. HYK02025 ML
Medchemexpress, HY-K0202 5 mL Protein A/G Magnetic Beads CAS: Protein A/G Magnetic Beads provide a fast and convenient method for Immunuoprecipitaion and Co-Immunoprecipitation and Chromatin Immunoprecipitation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.