Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein A High Performance
Catalog No. 502904836
Encompass
protein A HP SpinTrap(TM) are designed for the enrichment of proteins of interest immunoprecipitation and the purification of monoclonal and polyclonal antibodies from cell lysates and biological fluids. Contain protein A Sepharose(R) HP.
Catalog No. 50-290-4836 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28903132
protein A HP SpinTrap(TM) are designed for the enrichment of proteins of interest immunoprecipitation and the purification of monoclonal and polyclonal antibodies from cell lysates and biological fluids. Contain protein A Sepharose(R) HP.