Learn More
Medchemexpress LLC IgG-binding A protein (His) | 91932-65-9 | MFCD00081934 | 97.0% | 50UG
SDP
Catalog No. 5000176359
Encompass_Preferred
IgG-binding A protein (His) is a recombinant fragment of Staphylococcus aureus Protein A (residues N35-D330) expressed in Escherichia coli and supplied as a lyophilized, C-terminal 6xHis-tagged reagent. It binds the Fc region of IgG and is formulated for use in immunoassays, antibody capture, and assay development.
  • Binds the Fc region of IgG enabling antibody detection and capture.
  • C-terminal 6xHis tag facilitates purification and detection.
  • Lyophilized formulation for long-term storage and transport.
  • High purity (greater than 95%) supports sensitive immunoassays.
  • Low endotoxin (less than 1 EU/μg) suitable for many biological applications.
  • Stabilized formulation preserves activity after reconstitution.
Catalog No. 50-001-76359 Supplier Medchemexpress LLC Supplier No. HYP70032050UG
IgG-binding A protein (His) is a recombinant fragment of Staphylococcus aureus Protein A (residues N35-D330) expressed in Escherichia coli and supplied as a lyophilized, C-terminal 6xHis-tagged reagent. It binds the Fc region of IgG and is formulated for use in immunoassays, antibody capture, and assay development.
  • Binds the Fc region of IgG enabling antibody detection and capture.
  • C-terminal 6xHis tag facilitates purification and detection.
  • Lyophilized formulation for long-term storage and transport.
  • High purity (greater than 95%) supports sensitive immunoassays.
  • Low endotoxin (less than 1 EU/μg) suitable for many biological applications.
  • Stabilized formulation preserves activity after reconstitution.