Learn More
Bioworld Protein A, HRP conjugate, 1 mg
Supplier:  Bioworld 215600151
Encompass_Preferred
Chromatographically pure Protein A conjugated to horseradish peroxidase lyophilized from 0.02M Potassium Phosphate, 0.15M NaCl, pH 7.2, 10mg/ml BSA, and 0.01% Thimerosal.Key Applications:Dot Blot ELISA Western Blotting ImmunohistochemistryLyophilized: Stable for 2 years at -20°C from date of shipment. Rehydrated: Stable for 6 months at -20°C.For research use only. Not for human use or consumption.
Catalog No. 50-198-978