Learn More
Genscript Corporation Protein A MagBeads
Catalog No. NC1483387
Encompass
GenScript Protein A MagBeads are superparamagnetic beads covalently bound with recombinant protein A for antibody purification. The beads are supplied as 25% slurry in phosphate buffered saline (PBS), pH 7.4, containing 20% ethanol. The Protein A MagBeads have a binding capacity of more than 10 mg Rabbit IgG per 1 ml settled beads (e.g. 4 ml 25% slurry).
Catalog No. NC1483387 Supplier Genscript Corporation Supplier No. L00273
GenScript Protein A MagBeads are superparamagnetic beads covalently bound with recombinant protein A for antibody purification. The beads are supplied as 25% slurry in phosphate buffered saline (PBS), pH 7.4, containing 20% ethanol. The Protein A MagBeads have a binding capacity of more than 10 mg Rabbit IgG per 1 ml settled beads (e.g. 4 ml 25% slurry).