Learn More
Cell Signaling Technology Protein A Magnetic Beads, Immunoprecipitation, 5 mL
SDP
Catalog No. NC1404992
Encompass
These Protein A Magnetic Beads are an affinity matrix designed for the small-scale isolation of immunocomplexes from immunoprecipitation assays. The magnetic particles feature covalently coupled Protein A, which exhibits high affinity for IgG subclasses from various species, including human, rabbit, mouse, rat, and sheep. This system concentrates beads to the side of the tube for efficient separation, eliminating centrifugation steps, minimizing sample loss, and increasing washing efficiency.
  • Particle diameter of approximately 1.5 μm.
  • Binding capacity of > 0.2 μg Rabbit IgG/μl bead slurry.
  • Supplied in PBS with 0.1% BSA, 0.004% EDTA, and 0.075% sodium azide.
  • Compatible with standard magnetic separation racks to reduce processing time.
  • Designed for research use only.
Catalog No. NC1404992 Supplier Cell Signaling Technology Supplier No. 73778L
These Protein A Magnetic Beads are an affinity matrix designed for the small-scale isolation of immunocomplexes from immunoprecipitation assays. The magnetic particles feature covalently coupled Protein A, which exhibits high affinity for IgG subclasses from various species, including human, rabbit, mouse, rat, and sheep. This system concentrates beads to the side of the tube for efficient separation, eliminating centrifugation steps, minimizing sample loss, and increasing washing efficiency.
  • Particle diameter of approximately 1.5 μm.
  • Binding capacity of > 0.2 μg Rabbit IgG/μl bead slurry.
  • Supplied in PBS with 0.1% BSA, 0.004% EDTA, and 0.075% sodium azide.
  • Compatible with standard magnetic separation racks to reduce processing time.
  • Designed for research use only.