Learn More
Spherotech PROTEIN A POLYSTYRENE PARTICLE
Catalog No. NC1816707
Encompass_Preferred
Protein A Coated Polystyrene Particles.Concentration 0.5% w/v .Nominal size 6.0-8.0 µm5 mL
Catalog No. NC1816707 Supplier Spherotech Supplier No. PAP605
Protein A Coated Polystyrene Particles.Concentration 0.5% w/v .Nominal size 6.0-8.0 µm5 mL