Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Protein A, Recombinant

Catalog No. ICN987051 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN987051 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN987051 Supplier MP Biomedicals™ Supplier No. 08987051
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein A is useful for the study of cell surface antigens and receptors, and for detecting antibody-secreting cells.

  • Recombinant protein A binds immunoglobulins (primarily IgG) at the Fc region with high affinity.
  • In some cases, IgM and IgA will also bind. This property provides a rapid, simple and economical method for purification and analysis of a broad-spectrum of antibodies.
  • Fractionation of IgG subclasses can also be performed using recombinant protein A.
  • Because of its high affinity for immunoglobulins of many species and its low level of non-specific binding, recombinant protein A is useful for the study of cell surface antigens and receptors, and for detecting antibody-secreting cells.
  • Recombinant protein A is produced in an E. coli strain which contains the gene for mature protein A from Staphylococcus aureus Cowan I strain.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein A Sepharose
For Use With (Application) Immunoassays
Content And Storage 0°C to 5°C
Quantity 5 mg

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.