Learn More
Bioworld PROTEIN-A RED SEPAROPOREA
Supplier:  Bioworld 201810862
Encompass
PROTEIN-A RED SEPAROPORE (AGAROSE) 4B (MACROBEADS): Protein ARed Separopore 4B (MB) Affinity matrix with immobilized Protein-A by CNBr activation coupling method.; For purification and/or fractionation of IgG subclasses.; Reliable tool ...
Catalog No. 50-197-892