Learn More
Genscript Corporation Protein A Resin FF Prepacked C
Supplier:  Genscript Corporation L0068051
Encompass
GenScript Protein A Resin FF is an affinity chromatography media designed for easy one-step purification of classes subclasses and fragments of immunoglobulins from biological fluids and from cell culture media
Catalog No. 50-000-59057