Learn More
Bioworld PROTEIN-A SEPAROPORE 4B PR
Supplier:  Bioworld 201700211
Encompass
PROTEIN-A SEPAROPORE 4B, PREPACKED COLUMNS 2X 1ML: Protein A Separopore 4B Affinity matrix with immobilized Protein-A (from staphylococcus aureus) by CNBr activation coupling method. For purification and/or fractionation of IgG subclasse...
Catalog No. 50-197-639