Learn More
Bioworld PROTEIN-A SEPAROPORE 4B PR
Supplier:  Bioworld 201700221
Encompass
PROTEIN-A SEPAROPORE 4B, PREPACKED COLUMN 2X 2 ML: Protein A Separopore 4B Affinity matrix with immobilized Protein-A (from staphylococcus aureus) by CNBr activation coupling method. For purification and/or fractionation of IgG subclasse...
Catalog No. 50-197-640