Learn More
Bioworld PROTEIN-A SEPAROPOREAGARO
Supplier:  Bioworld 201810281
Encompass
PROTEIN-A SEPAROPORE (AGAROSE) (LYOPHILIZED) 4B-CL: Protein A Separopore 4B-CL Affinity matrix with immobilized Protein-A (from staphylococcus aureus) by CNBr activation coupling method.; For purification and/or fractionation of IgG sub...
Catalog No. 50-197-732