Learn More
Invitrogen™ Protein A - Sepharose™ 4B
Description
This product is supplied as a 50% suspension consisting of Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).
Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
Specifications
Specifications
| Product Type | Protein A Sepharose |
| Product Line | Sepharose |
| Column Type | Affinity |
| Stationary Phase | Protein A |
| Content And Storage | This product is supplied as a 50% suspension (5 ml Protein A-Sepharose™ 4B + 5 ml PBS) Wet Ice |
| Quantity | 5 mL |
| Form | Suspension |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.