Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein A-SepharoseR from250MG
Catalog No. 502904963
Encompass
Protein A-Sepharose when linked with antibody helps improving antigen binding capacity of IgG crosslinked to column matrices. In this case binding happens between Protein A and fc portion of the IgG molecule leaving antigen specific sites free.
Catalog No. 50-290-4963 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P3391250MG
Protein A-Sepharose when linked with antibody helps improving antigen binding capacity of IgG crosslinked to column matrices. In this case binding happens between Protein A and fc portion of the IgG molecule leaving antigen specific sites free.