Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein A SepharoseR from5G
Catalog No. 502914665
Encompass
Protein A-Sepharose when linked with antibody helps improving antigen binding capacity of IgG crosslinked to column matrices. In this case binding happens between Protein A and fc portion of the IgG molecule leaving antigen specific sites free.
Catalog No. 50-291-4665 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P33915G
Protein A-Sepharose when linked with antibody helps improving antigen binding capacity of IgG crosslinked to column matrices. In this case binding happens between Protein A and fc portion of the IgG molecule leaving antigen specific sites free.