Learn More
CUBE BIOTECH AChR T.Californica; 80 µg
Catalog No. NC2684617
Encompass
80 ¿g of AchR from T. Californica purified using cobra neuro-toxin3
Catalog No. NC2684617 Supplier CUBE BIOTECH Supplier No. 28603
80 ¿g of AchR from T. Californica purified using cobra neuro-toxin3