Learn More
Sino Biological Recombinant Aequorea victoria GFP Protein, Biotinylated 100 µg
Catalog No. 5000656700
Encompass_Preferred
Recombinant full-length Aequorea victoria GFP was expressed in E. coli cells using an N-terminal His-AVI tag, and biotinylated in vivo prior to affinity purification. The protein accession number is P42212.
Catalog No. 50-006-56700 Supplier Sino Biological Supplier No. E62030HB100
Recombinant full-length Aequorea victoria GFP was expressed in E. coli cells using an N-terminal His-AVI tag, and biotinylated in vivo prior to affinity purification. The protein accession number is P42212.