Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein AG from proprietar1MG
Catalog No. 502901790
Encompass
Protein G is a group G Streptococcus protein and is a large multi-domain cell wall protein. It interacts with the fc region of IgG (immunolglobulin) through its repeating 55-amino acid domain. Strain GX7809 and GX7805 contain two and three such protein repeats respectively.
Catalog No. 50-290-1790 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P46891MG
Protein G is a group G Streptococcus protein and is a large multi-domain cell wall protein. It interacts with the fc region of IgG (immunolglobulin) through its repeating 55-amino acid domain. Strain GX7809 and GX7805 contain two and three such protein repeats respectively.