Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein AG from proprietar5MG
Catalog No. 502904188
Encompass
Protein G is a group G Streptococcus protein and is a large multi-domain cell wall protein. It interacts with the fc region of IgG (immunolglobulin) through its repeating 55-amino acid domain. Strain GX7809 and GX7805 contain two and three such protein repeats respectively.
Catalog No. 50-290-4188 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P46895MG
Protein G is a group G Streptococcus protein and is a large multi-domain cell wall protein. It interacts with the fc region of IgG (immunolglobulin) through its repeating 55-amino acid domain. Strain GX7809 and GX7805 contain two and three such protein repeats respectively.