Learn More
Biochain Institute Inc Western Blot Enhancing Kit, 2500 cm2/PK
SDP
Catalog No. 50183003
Encompass_Preferred
BioChain's Attoglow Western Blot Enhancing Kit contains Millennium Enhancer which can significantly boost Western blot signals, and is compatible with other enhancing mechanisms to achieve multiplied enhancing effects.
Catalog No. 50-183-003 Supplier Biochain Institute Inc Supplier No. K3172250
BioChain's Attoglow Western Blot Enhancing Kit contains Millennium Enhancer which can significantly boost Western blot signals, and is compatible with other enhancing mechanisms to achieve multiplied enhancing effects.