Learn More
Biochain Institute Inc Attoglow Western Blot Analysis Kit: Millennium Enhancer, 100 ml/PK
SDP
Catalog No. 50182979
Encompass_Preferred
BioChain's Attoglow Western Blot Analysis Kit is an ultra-sensitive chemiluminescent Western blot assay. It detects protein at attogram (10-18 g) or yoctomole (10-24 M) levels. This kit consists of three parts: Millennium Enhancer, protein interaction system and detection system.
Catalog No. 50-182-979 Supplier Biochain Institute Inc Supplier No. K31712501
BioChain's Attoglow Western Blot Analysis Kit is an ultra-sensitive chemiluminescent Western blot assay. It detects protein at attogram (10-18 g) or yoctomole (10-24 M) levels. This kit consists of three parts: Millennium Enhancer, protein interaction system and detection system.