Learn More
Biochain Institute Inc Attoglow Western Blot Analysis Kit- with Millennium Enhancer, Anti chicken secondary antibody, 1200 cm2/PK
SDP
Catalog No. 50182977
Encompass_Preferred
Attoglow Western Blot Analysis Kit is an ultra-sensitive chemiluminescent Western blot analysis kit developed by BioChain. It detects protein at attogram (10-18 g) or yoctomole (10-24 M) levels. This kit consists of three parts: Millennium Enhancer, protein interaction system and detection system.
Catalog No. 50-182-977 Supplier Biochain Institute Inc Supplier No. K3171120III
Attoglow Western Blot Analysis Kit is an ultra-sensitive chemiluminescent Western blot analysis kit developed by BioChain. It detects protein at attogram (10-18 g) or yoctomole (10-24 M) levels. This kit consists of three parts: Millennium Enhancer, protein interaction system and detection system.