Learn More
Bio Rad Laboratories PROTEIN ASSAY DYE RGNT 450MLP
Catalog No. NC9279633
Encompass
Bio-Rad Protein Assay Dye Reagent Concentrate, 450mL, A colorimetric assay for measuring total protein concentration, The dye binds to primarily basic and aromatic amino acid residues
Catalog No. NC9279633 Supplier Bio Rad Laboratories Supplier No. 5000006
Bio-Rad Protein Assay Dye Reagent Concentrate, 450mL, A colorimetric assay for measuring total protein concentration, The dye binds to primarily basic and aromatic amino acid residues