Learn More
Leica Biosystems Protein Block 25ml
Supplier:  Leica Biosystems RE7102CE
Encompass
Novocastra Protein Block, RE7102, is intended for use in immunohistochemical (IHC) staining procedures. In immunohistochemistry, diffuse non-specific staining (background) may occur as a result of hydrophobic and ionic interactions between antibodies and tissue components. Novocastra Protein Block, RE7102, is a serum-free, protein blocker. 25 mL of reagent is supplied.
Catalog No. 50-255-1992