Learn More
Wl Gore And Associates 9.0 mL GORE Protein Capture Device with Protein A for Rapid mAb Purification
Catalog No. NC1931872
Encompass
GORE Protein Capture Devices with immobilized Protein A are intended for affinity purification of monoclonal antibodies and other proteins demonstrating an Fc region in process development or late phase discovery. GORE Protein Capture Devices use a unique expanded polytetrafluoroethylene (ePTFE) membrane composite that provides a binding capacity advantage at high flow rates and low pressure drop, and improved the speed of purification. In addition the GORE Protein Capture Deivces produce highly concentrated elution pools, which may eliminate a downstream concentration step during the antibody purification process. These unique membrane devices are offered in 1 mL (≥ 30 mg capacity), 3.5 mL (≥ 105 mg capacity) and 9.0 mL (≥27 mg capacity) at 20 seconds residence time.
Catalog No. NC1931872 Supplier Wl Gore And Associates Supplier No. PROA103
GORE Protein Capture Devices with immobilized Protein A are intended for affinity purification of monoclonal antibodies and other proteins demonstrating an Fc region in process development or late phase discovery. GORE Protein Capture Devices use a unique expanded polytetrafluoroethylene (ePTFE) membrane composite that provides a binding capacity advantage at high flow rates and low pressure drop, and improved the speed of purification. In addition the GORE Protein Capture Deivces produce highly concentrated elution pools, which may eliminate a downstream concentration step during the antibody purification process. These unique membrane devices are offered in 1 mL (≥ 30 mg capacity), 3.5 mL (≥ 105 mg capacity) and 9.0 mL (≥27 mg capacity) at 20 seconds residence time.