Learn More
Cayman Chemical Protein Carbonyl Assay Buffer, 60 mL
SDP
Catalog No. 502893490
Encompass_Preferred
Protein Carbonyl Assay Buffer has been tested and formulated to work exclusively with Cayman’s Protein Carbonyl Fluorometric Assay Kit
Catalog No. 50-289-3490 Supplier Cayman Chemical Supplier No. 70153260 ML
Protein Carbonyl Assay Buffer has been tested and formulated to work exclusively with Cayman’s Protein Carbonyl Fluorometric Assay Kit