Learn More
Apexbio Technology LLC PROTEIN CARBONYL COL KIT-96mg
SDP
Catalog No. 5000567703
Encompass_Preferred
Protein Carbonyl Colorimetric Assay Kit (Tissue and Serum Samples) is designed for the detection of protein oxidation commonly used in oxidative stress research Originating in biochemical laboratories this kit measures carbonyl groups marking oxidative protein damage via colorimetric detection mechanisms It targets pathways related to oxidative stress and aging In vitro the assay demonstrates efficiency in detecting protein carbonyls at a nanomolar to low micromolar range Typical applications include cell models for studying oxidative stress animal studies and drug screening for antioxidants Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67703 Supplier Apexbio Technology LLC Supplier No. K462496
Protein Carbonyl Colorimetric Assay Kit (Tissue and Serum Samples) is designed for the detection of protein oxidation commonly used in oxidative stress research Originating in biochemical laboratories this kit measures carbonyl groups marking oxidative protein damage via colorimetric detection mechanisms It targets pathways related to oxidative stress and aging In vitro the assay demonstrates efficiency in detecting protein carbonyls at a nanomolar to low micromolar range Typical applications include cell models for studying oxidative stress animal studies and drug screening for antioxidants Experimental concentrations typically depend on the specific research design