Learn More
Cayman Chemical Protein Carbonyl Fluorometric Assay Kit, 96 Well
SDP
Catalog No. 502892280
Encompass_Preferred
Cayman’s Protein Carbonyl Fluorometric Assay Kit utilizes the rhodamine B hydrazide (RBH) reaction to measure the protein carbonyl content in plasma, serum, cell lysates, or tissue homogenates in a convenient 96-well format. Formation of the fluorescent protein carbonyl-RBH hydrazone is analyzed using an excitation wavelength of 560 nm and an emission wavelength of 585-595 nm.
Catalog No. 50-289-2280 Supplier Cayman Chemical Supplier No. 70153096 WELL
Cayman’s Protein Carbonyl Fluorometric Assay Kit utilizes the rhodamine B hydrazide (RBH) reaction to measure the protein carbonyl content in plasma, serum, cell lysates, or tissue homogenates in a convenient 96-well format. Formation of the fluorescent protein carbonyl-RBH hydrazone is analyzed using an excitation wavelength of 560 nm and an emission wavelength of 585-595 nm.