Learn More
Apexbio Technology LLC PROTEIN CARBONYL KIT-100mg
SDP
Catalog No. 5000897383
Encompass_Preferred
Protein Carbonyl Content Assay Kit is an analytical tool used in the field of oxidative stress research It originates from the need to quantify carbonyl groups in proteins which arise due to oxidative damage The mechanism involves the derivatization of carbonyl groups to form stable hydrazones which can be measured spectrophotometrically This assay is routinely employed in vitro to assess protein oxidation levels in nanomolar to low micromolar ranges It is commonly applied in cell models animal studies and drug screening to evaluate oxidative stress markers Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97383 Supplier Apexbio Technology LLC Supplier No. K2188100
Protein Carbonyl Content Assay Kit is an analytical tool used in the field of oxidative stress research It originates from the need to quantify carbonyl groups in proteins which arise due to oxidative damage The mechanism involves the derivatization of carbonyl groups to form stable hydrazones which can be measured spectrophotometrically This assay is routinely employed in vitro to assess protein oxidation levels in nanomolar to low micromolar ranges It is commonly applied in cell models animal studies and drug screening to evaluate oxidative stress markers Experimental concentrations typically depend on the specific research design