Learn More
Sino Biological Protein CD70 ET-FREE 100UG
Catalog No. 5001073979
Encompass_Preferred
A DNA sequence encoding the Human CD70 (NP 001243 1) (Gln39-Pro193) was expressed with a polyhistidine tag at the N-terminus This construct has a trimer design
Catalog No. 50-010-73979 Supplier Sino Biological Supplier No. 10780H07H5UE100
A DNA sequence encoding the Human CD70 (NP 001243 1) (Gln39-Pro193) was expressed with a polyhistidine tag at the N-terminus This construct has a trimer design