Learn More
Sino Biological Protein CD9 ET-FREE 100UG
Catalog No. 5001073835
Encompass_Preferred
A DNA sequence encoding the second extracellular domain (Ser 112-Ile 195) of human CD9 (NP 001760 1) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus
Catalog No. 50-010-73835 Supplier Sino Biological Supplier No. 11029H08HUE100
A DNA sequence encoding the second extracellular domain (Ser 112-Ile 195) of human CD9 (NP 001760 1) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus