Learn More
Viva Products BJP-10/40 Clinical Concentrator
SDP
Supplier:  Viva Products BJP10/40
Encompass_Preferred
Clinical Ultrafiltration Concentrator; For concentration of proteins in clinical diagnostic samples such as urine and spinal fluid; Used prior to electrophoresis of monoclonal immunoglobulins (M-proteins) as well as immunofixation electrophoresis (IFE); Sample volume to 10 ml; Final concentrate volume as low as 50 microliters for 200X concentration factor; 7,500 MWCO; Number of tests is 40/PK
Catalog No. 50-355-739