Learn More
Viva Products BJP-5/100 Clinical Concentrator, with pipettes
SDP
Supplier:  Viva Products BJP5/100P
Encompass_Preferred
Clinical Ultrafiltration Concentrator; For concentration of proteins in clinical diagnostic samples such as urine and spinal fluid; Used prior to electrophoresis of monoclonal immunoglobulins (M-proteins) as well as immunofixation electrophoresis (IFE); Sample volume to 5 ml; Final concentrate volume as low as 50 microliters for 100X concentration factor; 7,500 MWCO; Includes disposable pipettes; Number of tests is 100/PK
Catalog No. 50-311-840