Learn More
Bioworld Protein-Coupling Buffer Kit, 1 Kit
Catalog No. 50198511
Encompass_Preferred
bioWORLD's Protein Coupling Buffer kit contains reagents/buffers necessary for convenient protein coupling to CNBr activated Sepharose or Separopore.Kit Components:Swell Buffer: 1L Coupling Buffer: 1L Wash buffer: 1LBlocking Buffer: 50mlEquilibrating Buffer: 100 mlReagents provided are sufficient for 25ml of gel.
Catalog No. 50-198-511 Supplier Bioworld Supplier No. 208700001
bioWORLD's Protein Coupling Buffer kit contains reagents/buffers necessary for convenient protein coupling to CNBr activated Sepharose or Separopore.Kit Components:Swell Buffer: 1L Coupling Buffer: 1L Wash buffer: 1LBlocking Buffer: 50mlEquilibrating Buffer: 100 mlReagents provided are sufficient for 25ml of gel.