Learn More
Sino Biological Protein Cyno EpCAM ET-FREE 1MG
Catalog No. 5001074168
Encompass_Preferred
A DNA sequence encoding the cynomolgus EPCAM (XP 005576740 1) (Met1-Lys265) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74168 Supplier Sino Biological Supplier No. 90299C08HUE1
A DNA sequence encoding the cynomolgus EPCAM (XP 005576740 1) (Met1-Lys265) was expressed with a polyhistidine tag at the C-terminus