Learn More
Sino Biological Protein Cyno ERBB2 Biotin 20UG
Catalog No. 5001073805
Encompass_Preferred
A DNA sequence encoding the Cynomolgus Her2/ERBB2 (XP 005584091 4) (Thr23-Thr652) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-73805 Supplier Sino Biological Supplier No. 90295C49H1B20
A DNA sequence encoding the Cynomolgus Her2/ERBB2 (XP 005584091 4) (Thr23-Thr652) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed