Learn More
Sino Biological Protein Cyno IL13 ET-FREE 50UG
Catalog No. 5001073696
Encompass_Preferred
A DNA sequence encoding the Cynomolgus IL-13 (ABG75889 1) (Met1-Asn132) was expressed with the Fc region of human IgG1 at the C-terminus
Catalog No. 50-010-73696 Supplier Sino Biological Supplier No. 11057C02HUE50
A DNA sequence encoding the Cynomolgus IL-13 (ABG75889 1) (Met1-Asn132) was expressed with the Fc region of human IgG1 at the C-terminus