Learn More
Sino Biological Protein Cyno LDLR ET-FREE 100
Catalog No. 5001074146
Encompass_Preferred
A DNA sequence encoding the Cynomolgus LDLR/LDL Receptor (XP 005588053 3) (Ala22-Gly788) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74146 Supplier Sino Biological Supplier No. 91058C08HUE100
A DNA sequence encoding the Cynomolgus LDLR/LDL Receptor (XP 005588053 3) (Ala22-Gly788) was expressed with a polyhistidine tag at the C-terminus