Learn More
Sino Biological Protein Cyno PDCD1 Biotin 100
Catalog No. 5001073803
Encompass_Preferred
A DNA sequence encoding the Cynomolgus PD-1 (B0LAJ3) (Met1-Gln167) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-73803 Supplier Sino Biological Supplier No. 90311C49HB100
A DNA sequence encoding the Cynomolgus PD-1 (B0LAJ3) (Met1-Gln167) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed