Learn More
Sino Biological Protein Cyno TNFSF15 100
Catalog No. 5000699121
Encompass_Preferred
A DNA sequence encoding the Cynomolgus/Rhesus TNFSF15 (F6S8F9) (Leu72-Leu251) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.
Catalog No. 50-006-99121 Supplier Sino Biological Supplier No. 91015C07H1UE100
A DNA sequence encoding the Cynomolgus/Rhesus TNFSF15 (F6S8F9) (Leu72-Leu251) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.