Learn More
Sino Biological Protein Cynomolgus ANP32B 1MG
Catalog No. 5001073907
Encompass_Preferred
A DNA sequence encoding the Cynomolgus APRIL/TNFSF13 (XP 005582818 1) (Gln111-Leu250) was fused with the Fc region of human IgG1 at the N-terminus This construct has a trimer design
Catalog No. 50-010-73907 Supplier Sino Biological Supplier No. 91072C01H1
A DNA sequence encoding the Cynomolgus APRIL/TNFSF13 (XP 005582818 1) (Gln111-Leu250) was fused with the Fc region of human IgG1 at the N-terminus This construct has a trimer design