Learn More
Sino Biological Protein Cynomolgus CD70 1MG
Catalog No. 5000699158
Encompass_Preferred
A DNA sequence encoding the Cynomolgus CD70/CD27 Ligand (XP_005587773.3) (Gln39-Pro194) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-006-99158 Supplier Sino Biological Supplier No. 91021C01HUE1
A DNA sequence encoding the Cynomolgus CD70/CD27 Ligand (XP_005587773.3) (Gln39-Pro194) was expressed with the Fc region of human IgG1 at the N-terminus.