Learn More
Sino Biological Protein Cynomolgus HHLA2 100UG
Catalog No. 5000699082
Encompass_Preferred
A DNA sequence encoding the Cynomolgus HHLA2 (XP_005548285.1) (Met1-Asn345) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-99082 Supplier Sino Biological Supplier No. 90982C08HUE100
A DNA sequence encoding the Cynomolgus HHLA2 (XP_005548285.1) (Met1-Asn345) was expressed with a polyhistidine tag at the C-terminus.