Learn More
Sino Biological Protein Cynomolgus HHLA2 1MG
Catalog No. 5000698804
Encompass_Preferred
A DNA sequence encoding the Cynomolgus HHLA2 (XP_005548285.1) (Met1-Asn345) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-98804 Supplier Sino Biological Supplier No. 90982C08HUE1
A DNA sequence encoding the Cynomolgus HHLA2 (XP_005548285.1) (Met1-Asn345) was expressed with a polyhistidine tag at the C-terminus.