Learn More
Sino Biological Recombinant Cynomolgus Integrin alpha 4 beta 7 Protein (His Tag), HPLC-verified 100 µg
Catalog No. 5000656668
Encompass_Preferred
A DNA sequence encoding the Cynomolgus ITGA4 (XP_005573683.3)(Glu20-Glu597) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1 ; A DNA sequence encoding the Cynomolgus ITGB7 (XP_005571028.3)(Tyr34-Lys771) was expressed, constructed the plasmid 2. The two plasmids were co-expressed and the Cynomolgus Integrin alpha 4 beta 7 Heterodimer was purified.
Catalog No. 50-006-56668 Supplier Sino Biological Supplier No. CT171C08H2100
A DNA sequence encoding the Cynomolgus ITGA4 (XP_005573683.3)(Glu20-Glu597) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1 ; A DNA sequence encoding the Cynomolgus ITGB7 (XP_005571028.3)(Tyr34-Lys771) was expressed, constructed the plasmid 2. The two plasmids were co-expressed and the Cynomolgus Integrin alpha 4 beta 7 Heterodimer was purified.